0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,6-Diethylaniline
BP0000757150
2,6-Diethylaniline
New Product
:
Apollo Scientific
2,6-Diethylaniline
:
Apollo Scientific
New Product
2,6-Diethylaniline
Product Family Options
Pack type
APOSOR93652-25G
Bottle
1 * 25 g
1 * 25 g - 21,80 €
21,80 €
-
+
1 * 25 g
21,80 €
-
+
New Product
New Product
APOSOR93652-100G
Bottle
1 * 100 g
1 * 100 g - 24,60 €
24,60 €
-
+
1 * 100 g
24,60 €
-
+
New Product
New Product
APOSOR93652-500G
Bottle
1 * 500 g
1 * 500 g - 57,90 €
57,90 €
-
+
1 * 500 g
57,90 €
-
+
New Product
New Product