0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Theacrine
BP0000755607
Tetramethyluric acid
New Product
:
Apollo Scientific
Tetramethyluric acid
:
Apollo Scientific
New Product
Tetramethyluric acid
Product Family Options
Pack type
APOSOR75078-25G
Bottle
1 * 25 g
1 * 25 g - 42,00 €
42,00 €
-
+
1 * 25 g
42,00 €
-
+
New Product
New Product
APOSOR75078-500G
Bottle
1 * 500 g
1 * 500 g - 728,00 €
728,00 €
-
+
1 * 500 g
728,00 €
-
+
New Product
New Product
APOSOR75078-100G
Bottle
1 * 100 g
1 * 100 g - 275,00 €
275,00 €
-
+
1 * 100 g
275,00 €
-
+
New Product
New Product
APOSOR75078-1G
Bottle
1 * 1 g
1 * 1 g - 71,00 €
71,00 €
-
+
1 * 1 g
71,00 €
-
+
New Product
New Product
APOSOR75078-5G
Bottle
1 * 5 g
1 * 5 g - 73,80 €
73,80 €
-
+
1 * 5 g
73,80 €
-
+
New Product
New Product