Terbumeton
Terbumeton
Catalog # EHERC17290000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCNc1nc(NC(C)(C)C)nc(OC)n1 Inchi: InChI=1S/C10H19N5O/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15) IUPAC: 4-N-tert-butyl-2-N-ethyl-6-methoxy-1,3,5-triazine-2,4-diamine
- Packing group: III
Specifications
Specifications
- Description:Terbumeton
- UN No.:3077
- Cat. No.:EHERC17290000
- Formula:C₁₀H₁₉N₅O
- Storage conditions:20 ℃
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Terbumeton; 1,3,5-Triazine-2,4-diamine, N2-(1,1-dimethylethyl)-N4-ethyl-6-methoxy- (ACI); 1,3,5-Triazine-2,4-diamine, N-(1,1-dimethylethyl)-N'-ethyl-6-methoxy- (9CI); N2-(1,1-Dimethylethyl)-N4-ethyl-6-methoxy-1,3,5-triazine-2,4-diamine (ACI); s-Triazine, 2-(tert-butylamino)-4-(ethylamino)-6-methoxy- (8CI); 2-Methoxy-4-ethylamino-6-tert-butylamino-s-triazine; 2-N-tert-Butyl-4-N-ethyl-6-methoxy-1,3,5-triazine-2,4-diamine; 4-(Ethylamino)-2-methoxy-6-(tert-butylamino)-s-triazine; A 2591; Caragard; GS 14259; Karagard; MEBT; Terbumeton; Terbuthylon; Terbutone; tert-Bumeton; Tribunil G 14259
- Molecular weight:225,29
- CAS No.:33693-04-8
- Shipping conditions:RT



