Strychnine
Strychnine
Catalog # EHERC16980000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1c7ccccc67 Inchi: InChI=1S/C21H22N2O2/c24-18-10-16-19-13-9-17-21(6-7-22(17)11-12(13)5-8-25-16)14-3-1-2-4-15(14)23(18)20(19)21/h1-5,13,16-17,19-20H,6-11H2/t13-,16-,17-,19-,20-,21+/m0/s1 IUPAC: (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
- Packing group: I
Specifications
Specifications
- Description:Strychnine
- UN No.:1692
- Cat. No.:EHERC16980000
- Formula:C₂₁H₂₂N₂O₂
- Storage conditions:20 ℃
- Hazard class description:GHS06, GHS09
- GHS signal:Danger
- Synonym(s):Strychnidin-10-one; Strychnine (8CI); 4,6-Methano-6H,14H-indolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline, strychnidin-10-one deriv.; (-)-Strychnine; Certox; NSC 5365; Strychnin
- Molecular weight:334,41
- CAS No.:57-24-9
- Shipping conditions:RT



