HHCB
HHCB
EHERC14213000
- SMILES: CC1OCCc2cc3c(cc12)C(C)(C)C(C)C3(C)C Inchi: InChI=1S/C18H26O/c1-11-9-19-10-13-7-15-16(8-14(11)13)18(5,6)12(2)17(15,3)4/h7-8,11-12H,9-10H2,1-6H3 IUPAC: 4,6,6,7,8,8-hexamethyl-1,3,4,7-tetrahydrocyclopenta[g]isochromene
- Packing group: III
- Description:HHCB
- UN No.:3082
- Cat. No.:EHERC14213000
- Formula:C₁₈H₂₆O
- Storage conditions:–18 ℃
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran; 1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyrane; 1,3,4,6,7,8-Hexahydro-4,6,6,8,8,8-hexamethylcyclopenta-2-benzopyran; 4,6,6,7,8,8-Hexamethyl-1,3,4,6,7,8-hexahydrocyclopenta[g]isochromene; Abbalide; Galaxolide; Galaxolide 50; Galaxolide 50BB; Galaxolide 50IPM; Galaxolide White; HHCB; Pearlide; Cyclopenta[g]-2-benzopyran, 1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethyl-
- Molecular weight:258,4
- CAS No.:1222-05-5
- Shipping conditions:RT



