Hexachlorophen
Hexachlorophen
Catalog # EHERC14180000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1c(Cl)cc(Cl)c(Cl)c1Cc2c(O)c(Cl)cc(Cl)c2Cl Inchi: InChI=1S/C13H6Cl6O2/c14-6-2-8(16)12(20)4(10(6)18)1-5-11(19)7(15)3-9(17)13(5)21/h2-3,20-21H,1H2 IUPAC: 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)methyl]phenol
- Packing group: III
Specifications
Specifications
- Description:Hexachlorophen
- UN No.:2875
- Cat. No.:EHERC14180000
- Formula:C₁₃H₆Cl₆O₂
- Storage conditions:20 ℃
- Hazard class description:GHS06, GHS09
- GHS signal:Danger
- Synonym(s):2,2',3,3',5,5'-Hexachloro-6,6'-dihydroxydiphenylmethane; 2,2'-Dihydroxy-3,3',5,5',6,6'-hexachlorodiphenylmethane; 2,2'-Dihydroxy-3,5,6,3',5',6'-hexachlorodiphenylmethane; 2,2'-Methylenebis[3,4,6-trichlorophenol]; AT 7; Acigena; Almederm; B 32; Bilevon; Bis(2-hydroxy-3,5,6-trichlorophenyl)methane; Bis(3,5,6-trichloro-2-hydroxyphenyl)methane; Cotofilm; Dermadex; Distodin; Exofene; Fesia-sin; Fostril; G 11; G 11 (bactericide); Gamophen; Gamophene; Germa-Medica; Hexabalm; Hexachlorofen; Hexachlorophen; Hexachlorophene; Hexafen; Hexophene; Hexosan; NSC 49115; NSC 9887; Nabac; Neosept V; Ritosept; Septisol; Septofen; Soy-Dome; Steral; Steraskin; Surgi-Cen; Surofene; Tersaseptic; pHisoHex; Phenol, 2,2'-methylenebis[3,4,6-trichloro-
- Molecular weight:406,9
- CAS No.:70-30-4
- Shipping conditions:RT



