Flufenzine
Flufenzine
Catalog # EHERC13715000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Fc1cccc(F)c1c2nnc(nn2)c3ccccc3Cl Inchi: InChI=1S/C14H7ClF2N4/c15-9-5-2-1-4-8(9)13-18-20-14(21-19-13)12-10(16)6-3-7-11(12)17/h1-7H IUPAC: 3-(2-chlorophenyl)-6-(2,6-difluorophenyl)-1,2,4,5-tetrazine
- Packing group: III
Specifications
Specifications
- Description:Flufenzine
- UN No.:3077
- Cat. No.:EHERC13715000
- Formula:C₁₄H₇ClF₂N₄
- Storage conditions:20 ℃
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):3-(2-Chlorophenyl)-6-(2,6-difluorophenyl)-1,2,4,5-tetrazine; Flufenzin; Flufenzine; Flumite 200; SZI 121
- Molecular weight:304,68
- CAS No.:162320-67-4
- Shipping conditions:RT



