Ethidimuron
Ethidimuron
Catalog # EHERC13240000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCS(=O)(=O)c1nnc(s1)N(C)C(=O)NC Inchi: InChI=1S/C7H12N4O3S2/c1-4-16(13,14)7-10-9-6(15-7)11(3)5(12)8-2/h4H2,1-3H3,(H,8,12) IUPAC: 1-(5-ethylsulfonyl-1,3,4-thiadiazol-2-yl)-1,3-dimethylurea
- Packing group: III
Specifications
Specifications
- Description:Ethidimuron
- UN No.:3077
- Cat. No.:EHERC13240000
- Formula:C₇H₁₂N₄O₃S₂
- Storage conditions:20 ℃
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Urea, 1-[5-(ethylsulfonyl)-1,3,4-thiadiazol-2-yl]-1,3-dimethyl- (8CI); N-[5-(Ethylsulfonyl)-1,3,4-thiadiazol-2-yl]-N,N'-dimethylurea; 1-(5-Ethylsulfonyl-1,3,4-thiadiazol-2-yl)-1,3-dimethylurea; BAY-MET 1486; Ethidimuron; MET 1486; Sulfodiazol; Ustilan; Urea, N-[5-(ethylsulfonyl)-1,3,4-thiadiazol-2-yl]-N,N'-dimethyl-
- Molecular weight:264,33
- CAS No.:30043-49-3
- Shipping conditions:RT



