Eprinomectin
Eprinomectin
EHERCA13187000
- SMILES: CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@H](NC(=O)C)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C.CO[C@H]8C[C@H](O[C@H]9[C@H](C)O[C@H](C[C@@H]9OC)O[C@H]%10[C@@H](C)\C=C\C=C\%11/CO[C@@H]%12[C@H](O)C(=C[C@@H](C(=O)O[C@H]%13C[C@@H](C\C=C\%10/C)O[C@@]%14(C%13)O[C@H](C(C)C)[C@@H](C)C=C%14)[C@]%11%12O)C)O[C@@H](C)[C@H]8NC(=O)C Inchi: InChI=1S/C50H75NO14.C49H73NO14/c1-12-26(2)45-29(5)18-19-49(65-45)24-36-21-35(64-49)17-16-28(4)44(27(3)14-13-15-34-25-58-47-43(53)30(6)20-37(48(54)61-36)50(34,47)55)62-41-23-39(57-11)46(32(8)60-41)63-40-22-38(56-10)42(31(7)59-40)51-33(9)52;1-25(2)43-28(5)17-18-48(64-43)23-35-20-34(63-48)16-15-27(4)44(26(3)13-12-14-33-24-57-46-42(52)29(6)19-36(47(53)60-35)49(33,46)54)61-40-22-38(56-11)45(31(8)59-40)62-39-21-37(55-10)41(30(7)58-39)50-32(9)51/h13-16,18-20,26-27,29,31-32,35-47,53,55H,12,17,21-25H2,1-11H3,(H,51,52);12-15,17-19,25-26,28,30-31,34-46,52,54H,16,20-24H2,1-11H3,(H,50,51)/b14-13+,28-16+,34-15+;13-12+,27-15+,33-14+/t26-,27-,29-,31-,32-,35+,36-,37-,38-,39-,40-,41-,42+,43+,44-,45+,46-,47+,49+,50+;26-,28-,30-,31-,34+,35-,36-,37-,38-,39-,40-,41+,42+,43+,44-,45-,46+,48+,49+/m00/s1
- Packing group: II
- Description:Eprinomectin
- UN No.:2811
- Cat. No.:EHERCA13187000
- Formula:C₅₀H₇₅NO₁₄·C₄₉H₇₃NO₁₄
- Storage conditions:–18 ℃
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):(4''R)-4''-(Acetylamino)-4''-deoxyavermectin B1; Eprinex; Eprinomectin; Ivomec Eprinex; MK 397
- Molecular weight:1814,23
- CAS No.:123997-26-2
- Shipping conditions:RT



