Camphor
Camphor
Catalog # EHERC10945400
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2 Inchi: InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1 IUPAC: (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
- Packing group: III
Specifications
Specifications
- Description:Camphor
- UN No.:2717
- Cat. No.:EHERC10945400
- Formula:C₁₀H₁₆O
- Storage conditions:4 ℃
- Hazard class description:GHS02, GHS05, GHS07, GHS08
- GHS signal:Warning
- Synonym(s):Camphor racemic; (1RS,4RS)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one; Camphor; 1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone; 1,7,7-Trimethylnorcamphor; 2-Bornanone; 2-Camphanone; Alphanon; Borneo camphor; DL-Camphor; Root bark oil; Spirit of camphor; dl-Camphor; (+/-)-Camphor
- Molecular weight:152,23
- CAS No.:76-22-2
- Shipping conditions:RT



