Bromobenzene
Bromobenzene
Catalog # EHERCA10710000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Brc1ccccc1 Inchi: InChI=1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H IUPAC: bromobenzene
- Packing group: III
Specifications
Specifications
- Description:Bromobenzene
- UN No.:2514
- Cat. No.:EHERCA10710000
- Formula:C₆H₅Br
- Storage conditions:20 ℃
- Hazard class description:GHS02, GHS07, GHS09
- GHS signal:Warning
- Synonym(s):1-Bromobenzene; Bromobenzene; Bromobenzol; Monobromobenzene; NSC 6529; Phenyl bromide
- Molecular weight:157,01
- CAS No.:108-86-1
- Shipping conditions:RT



