Benzoximate
Benzoximate
Catalog # EHERC10540000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCON=C(OC(=O)c1ccccc1)c2c(OC)ccc(Cl)c2OC Inchi: InChI=1S/C18H18ClNO5/c1-4-24-20-17(25-18(21)12-8-6-5-7-9-12)15-14(22-2)11-10-13(19)16(15)23-3/h5-11H,4H2,1-3H3 IUPAC: [C-(3-chloro-2,6-dimethoxyphenyl)-N-ethoxycarbonimidoyl] benzoate
Specifications
Specifications
- Description:Benzoximate
- Cat. No.:EHERC10540000
- Formula:C₁₈H₁₈ClNO₅
- Storage conditions:20 ℃
- Hazard class description:GHS09
- GHS signal:Warning
- Molecular weight:363,79
- CAS No.:29104-30-1
- Shipping conditions:RT



