Anthanthrene
Anthanthrene
EHERC20515000
Jump to:
- SMILES: c1cc2ccc3cc4cccc5ccc6cc(c1)c2c3c6c45 Inchi: InChI=1S/C22H12/c1-3-13-7-9-18-12-16-6-2-4-14-8-10-17-11-15(5-1)19(13)21(18)22(17)20(14)16/h1-12H IUPAC: Anthanthrene
Specifications
Specifications
- Description:Anthanthrene
- Cat. No.:EHERC20515000
- Formula:C₂₂H₁₂
- Storage conditions:20 ℃
- Hazard class description:GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Dibenzo[def,mno]chrysene; Anthanthren; Anthanthrene; Dibenzo[cd,jk]pyrene
- Molecular weight:276,33
- CAS No.:191-26-4
- Shipping conditions:RT



