4-Nitroimidazole
4-Nitroimidazole
Catalog # EHERC15588300
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: [O-][N+](=O)c1c[nH]cn1 Inchi: InChI=1S/C3H3N3O2/c7-6(8)3-1-4-2-5-3/h1-2H,(H,4,5) IUPAC: 4-nitro-1H-imidazole
Specifications
Specifications
- Description:4-Nitroimidazole
- Cat. No.:EHERC15588300
- Formula:C₃H₃N₃O₂
- Storage conditions:20 ℃
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Metronidazole Imp. B (EP); 5-Nitro-1H-imidazole; 4-Nitro-1H-imidazole; 4(or 5)-Nitroimidazole; 1H-4-Nitroimidazole; 1H-5-Nitroimidazole; 4(5)-Nitroimidazole; 4-Nitroimidazole; 5-Nitroimidazole; NSC 50359; Metronidazole Impurity B
- Molecular weight:113,07
- CAS No.:3034-38-6
- Shipping conditions:RT



