3-Hexanol
3-Hexanol
Catalog # EHERC14196900
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCC(O)CC Inchi: InChI=1S/C6H14O/c1-3-5-6(7)4-2/h6-7H,3-5H2,1-2H3 IUPAC: hexan-3-ol
- Packing group: III
Specifications
Specifications
- Description:3-Hexanol
- UN No.:2282
- Cat. No.:EHERC14196900
- Formula:C₆H₁₄O
- Storage conditions:20 ℃
- Hazard class description:GHS02
- GHS signal:Warning
- Synonym(s):Hexan-3-ol; 3-Hexanol
- Molecular weight:102,17
- CAS No.:623-37-0
- Shipping conditions:RT



