2-Methylpentane
2-Methylpentane
Catalog # EHERC15121200
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCC(C)C Inchi: InChI=1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3 IUPAC: 2-methylpentane
- Packing group: II
Specifications
Specifications
- Description:2-Methylpentane
- UN No.:1208
- Cat. No.:EHERC15121200
- Formula:C₆H₁₄
- Storage conditions:20 ℃
- Hazard class description:GHS02, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Pentane, 2-methyl-; 2-Methylpentane; 1,1-Dimethylbutane; Isohexane; Kyowasol C 600M; Kyowazol C 600; NSC 66496
- Molecular weight:86,18
- CAS No.:107-83-5
- Shipping conditions:RT



