2,4-Dinitroaniline
2,4-Dinitroaniline
Catalog # EHERC12782000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Nc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-] Inchi: InChI=1S/C6H5N3O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H,7H2 IUPAC: 2,4-dinitroaniline
- Packing group: II
Specifications
Specifications
- Description:2,4-Dinitroaniline
- UN No.:1596
- Cat. No.:EHERC12782000
- Formula:C₆H₅N₃O₄
- Storage conditions:20 ℃
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):2,4-Dinitrobenzenamine; 1-Amino-2,4-dinitrobenzene; 2,4-Dinitroaniline; Dinitroaniline; NSC 8731
- Molecular weight:183,12
- CAS No.:97-02-9
- Shipping conditions:RT



