2,4-DB
2,4-DB
Catalog # EHERC11980000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC(=O)CCCOc1ccc(Cl)cc1Cl Inchi: InChI=1S/C10H10Cl2O3/c11-7-3-4-9(8(12)6-7)15-5-1-2-10(13)14/h3-4,6H,1-2,5H2,(H,13,14) IUPAC: 4-(2,4-dichlorophenoxy)butanoic acid
- Packing group: III
Specifications
Specifications
- Description:2,4-DB
- UN No.:3077
- Cat. No.:EHERC11980000
- Formula:C₁₀H₁₀Cl₂O₃
- Storage conditions:20 ℃
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Butanoic acid, 4-(2,4-dichlorophenoxy)-; Butyric acid, 4-(2,4-dichlorophenoxy)- (8CI); 4-(2,4-Dichlorophenoxy)butanoic acid; 2,4-DB; 2,4-DM; 2,4-Dichlorophenoxybutyric acid; 4-(2,4-Dichlorophenoxy)butyric acid; Buratal; Butyrac; Butyrac 118; Butyrac 200; Legumex; Legumex D; NSC 70337; Sys 67 Buratal; γ-(2,4-Dichlorophenoxy)butyric acid
- Molecular weight:249,09
- CAS No.:94-82-6
- Shipping conditions:Room temperature



