2,2-Dichloroethanol
2,2-Dichloroethanol
Catalog # EHERC12422350
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OCC(Cl)Cl Inchi: InChI=1S/C2H4Cl2O/c3-2(4)1-5/h2,5H,1H2 IUPAC: 2,2-dichloroethanol
Specifications
Specifications
- Description:2,2-Dichloroethanol
- Cat. No.:EHERC12422350
- Formula:C₂H₄Cl₂O
- Storage conditions:20 ℃
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):2,2-Dichloroethanol; Dichloroethanol; NSC 60513
- Molecular weight:114,96
- CAS No.:598-38-9
- Shipping conditions:RT



