Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,5-Pentanediol

1,5-Pentanediol
  EHERC15977700
1,5-Pentanediol
  EHERC15977700
 :  
1,5-Pentanediol
1,5-Pentanediol

1,5-Pentanediol

 :  
 
43,80 €
   
1 * 0,25 g
 
  • SMILES: OCCCCCO Inchi: InChI=1S/C5H12O2/c6-4-2-1-3-5-7/h6-7H,1-5H2 IUPAC: pentane-1,5-diol
 

 

  • Description:
    1,5-Pentanediol
  • Cat. No.:
    EHERC15977700
  • Formula:
    C₅H₁₂O₂
  • Storage conditions:
    20 ℃
  • Hazard class description:
    GHS07
  • GHS signal:
    Warning
  • Synonym(s):
    1,5-Pentanediol (8CI, 9CI, ACI); 1,5-Dihydroxypentane; 1,5-Pentamethylene glycol; Cosphaderm Pentiol natural; Dermosoft Pentiol Eco; Hydrolite 5 Green; Microcare Emollient PTG; NSC 5927; Pentamethylene glycol; Pentylene glycol; Purolan PD-LO; TK 5; α,ω-Pentanediol; ω-Pentanediol
  • Molecular weight:
    104,15
  • CAS No.:
    111-29-5
  • Shipping conditions:
    RT