Pentane-1,5-dithiol 98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
Synonyms: 1,5-Dimercaptopentane
- CAS Number:928-98-3
- Formula:C₅H₁₂S₂
- MDL Number:MFCD00004908
- Molecular Weight:136,27877 g/mol
4 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 1,5-Dimercaptopentane
- CAS Number:928-98-3
- Formula:C₅H₁₂S₂
- MDL Number:MFCD00004908
- Molecular Weight:136,27877 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



