Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxyquinazoline 98%

4-Hydroxyquinazoline 98%

Irritant
Supplier:   Apollo Scientific
4-Hydroxyquinazoline 98%
Supplier:  Apollo Scientific
Certificates
4-Hydroxyquinazoline 98%

6 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  4-Hydroxyquinazoline, Quinazolin-4(3H)-one
  • CAS Number:491-36-1
  • Formula:C₈H₆N₂O
  • MDL Number:MFCD00511302
  • Molecular Weight:146,14604 g/mol
  • EINECS:207-735-8
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 24,60 €
    24,60 €
    1 * 25 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 60,80 €
    60,80 €
    1 * 100 g
       60,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 157,00 €
    157,00 €
    1 * 500 g
       157,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 498,00 €
    498,00 €
    1 * 2,5 kg
       498,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 990,00 €
    1 990,00 €
    1 * 10 kg
       1 990,00 €

    Specifications:
    New Product
    New Product