Xylenes (total) 2000 µg/ml in Methanol
Xylenes (total) 2000 µg/ml in Methanol
Catalog # EHERGA09011025ME
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1=CC=CC(C)=C1.CC2=CC=CC=C2C.CC3=CC=C(C)C=C3
- Inchi: InChI=1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3
- IUPAC: m-xylene--o-xylene--p-xylene (1/1/1)
- Packing group: II
Specifications
Specifications
- Description:Xylenes (total) 2000 µg/ml in Methanol
- UN No.:1230M
- Cat. No.:EHERGA09011025ME
- Formula:C₈H₁₀
- Storage conditions:–18 °C
- Hazard class description:GHS02, GHS06, GHS08
- GHS signal:Danger
- Synonym(s):Dimethylbenzene;Entellan New;Xylol;ZEP-RD;
- Molecular weight:106,16
- CAS No.:1330-20-7
- Shipping conditions:RT



