Tri-allate
Tri-allate
Catalog # EHERC17630000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)N(C(C)C)C(=O)SCC(=C(Cl)Cl)Cl
- Inchi: InChI=1S/C10H16Cl3NOS/c1-6(2)14(7(3)4)10(15)16-5-8(11)9(12)13/h6-7H,5H2,1-4H3
- IUPAC: S-(2,3,3-trichloroprop-2-enyl) N,N-di(propan-2-yl)carbamothioate
- Packing group: III
Specifications
Specifications
- Description:Tri-allate
- UN No.:3077
- Cat. No.:EHERC17630000
- Formula:C₁₀H₁₆Cl₃NOS
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Synonym(s):2-Propene-1-thiol;2,3,3-trichloro-;diisopropylcarbamate (8CI);Carbamic acid;diisopropylthio-;S-(2,3,3-trichloroallyl) ester (7CI;8CI);Carbamothioic acid;bis(1-methylethyl)-;S-(2,3,3-trichloro-2-propenyl) ester (9CI);2,3,3-Trichloroallyl N,N-diisopropylthiocarbamate;Avadex BW;CP 23426;Dipthal;Far-Go;MeSH ID: D014214;NSC 379698;S-2,3,3-Trichloroallyl diisopropylthiocarbamate;S-2,3,3-Trichloroallyl N,N-diisopropylthiocarbamate;Triallate;Triamyl;Trillate
- Molecular weight:304,66
- CAS No.:2303-17-5
- Shipping conditions:RT



