Sudan Black B
Sudan Black B
Catalog # EHERC16986110
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1(C)Nc2cccc3c(ccc(N1)c23)N=Nc4ccc(N=Nc5ccccc5)c6ccccc46
- Inchi: InChI=1S/C29H24N6/c1-29(2)30-26-14-8-13-22-25(17-18-27(31-29)28(22)26)35-34-24-16-15-23(20-11-6-7-12-21(20)24)33-32-19-9-4-3-5-10-19/h3-18,30-31H,1-2H3/b33-32+,35-34+
- IUPAC: (E)-(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-[4-[(E)-phenyldiazenyl]naphthalen-1-yl]diazene
Specifications
Specifications
- Description:Sudan Black B
- Cat. No.:EHERC16986110
- Formula:C₂₉H₂₄N₆
- Storage conditions:20 °C
- Hazard class description:GHS08
- GHS signal:Danger
- Synonym(s):1H-Perimidine;2,3-dihydro-2,2-dimethyl-6-[2-[4-(2-phenyldiazenyl)-1-naphthalenyl]diazenyl]- (ACI);1H-Perimidine;2,3-dihydro-2,2-dimethyl-6-[[4-(phenylazo)-1-naphthalenyl]azo]- (9CI);2,3-Dihydro-2,2-dimethyl-6-[2-[4-(2-phenyldiazenyl)-1-naphthalenyl]diazenyl]-1H-perimidine (ACI);C.I. Solvent Black 3 (7CI;8CI);Sudan Black B (6CI);(2,2-Dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene;2,2-Dimethyl-6-[[4-(phenyldiazenyl)-1-naphthyl]diazenyl]-2,3-dihydro-1H-perimidine;2,3-Dihydro-2,2-dimethyl-6-[(4-phenylazo-1-naphthalenyl)-azo]-1H-perimidine;2,3-Dihydro-2,2-dimethyl-6-[[4-(phenylazo)-1-naphthyl]azo]perimidine;6-(1-Phenylazo-4-naphthylazo)-2,3-dihydro-2,2-dimethylperimidine;Aizen SOT Black 6;C.I. 26150;Ceres Black BN;Chuo Sudan Black 141;Fast Black HBN;Fat Black HB;Fat Black HB 01;Hexatype Black B;Lacquer Black S;Lacquer Black VB;Neptune Black X 60;NSC 11239;Nubian Black BT;Oil Black 141;Oil Black 860;Oil Black BT;Oil Black HBB;Oil Black HZ;Orient Oil Black 860;Orient Oil Black HBB;Polysolve Black 3;Solvent Black 3;SOT-Black 6;Sudan Black;Sudan Black 141;Sudan Black BB;Sudan Black X 60;Sudan Deep Black BB;Sudan Deep Black BN;Typogen Black
- Molecular weight:456,54
- CAS No.:4197-25-5
- Shipping conditions:RT



