Streptomycin sulfate
Streptomycin sulfate
EHERC16974900
- SMILES: CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]3NC(=N)N)O[C@@H](C)[C@]2(O)C=O.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@H](O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]6NC(=N)N)O[C@@H](C)[C@]5(O)C=O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
- Inchi: InChI=1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7+,8-,9-,10-,11+,12-,13-,14+,15+,16-,17-,18-,21+;;;/m00.../s1
- IUPAC: 1-[(1R,2R,3S,4R,5R,6S)-3-carbamimidamido-4-[(2R,3R,4R,5S)-3-[(2S,3S,4S,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;sulfuric acid
- Description:Streptomycin sulfate
- Cat. No.:EHERC16974900
- Formula:2C₂₁H₃₉N₇O₁₂·₃H₂O₄S
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Streptomycin sulfate;bis[N,N'-bis(aminoiminomethyl)-4-O-[5-deoxy-2-O-[2-deoxy-2-(methylamino)-a-l-glucopyranosyl]-3-C-formyl-a-l-lyxofuranosyl]-d-streptamine] trisulfate;Streptomycin;sulfate (2:3) (salt) (8CI);Ag Streptomycin;Agri-Strep;Agrimycin 17;Agrimycin sulfate;Ambistryn;Ambistryn S;FireWall (pesticide);Hortocyna;KeyStrepto;Plantomycin;Strepto;Streptobrettin;Streptomycin A sulfate;Streptomycin sesquisulfate;Streptomycin sesquisulphate;Streptomycin sulphate;Streptonex;Strycin;Vetstrep;D-Streptamine;O-2-deoxy-2-(methylamino)-alpha-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-formyl-alpha-L-lyxofuranosyl-(1→4)-N1,N3-bis(aminoiminomethyl)-;sulfate (2:3);D-Streptamine;O-2-deoxy-2-(methylamino)-alpha-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-formyl-alpha-L-lyxofuranosyl-(1→4)-N,N'-bis(aminoiminomethyl)-;sulfate (2:3) (salt) (9CI)
- Molecular weight:1457,38
- CAS No.:3810-74-0
- Shipping conditions:RT



