Propachlor
Propachlor
Catalog # EHERC16380000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)N(C(=O)CCl)c1ccccc1
- Inchi: InChI=1S/C11H14ClNO/c1-9(2)13(11(14)8-12)10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3
- IUPAC: 2-chloro-N-phenyl-N-propan-2-ylacetamide
- Packing group: III
Specifications
Specifications
- Description:Propachlor
- UN No.:2811
- Cat. No.:EHERC16380000
- Formula:C₁₁H₁₄ClNO
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS07, GHS09
- GHS signal:Danger
- Synonym(s):Acetanilide;2-chloro-N-isopropyl- (8CI);2-Chloro-N-(1-methylethyl)-N-phenylacetamide;2-Chloro-N-isopropyl-N-phenylacetamide;2-Chloro-N-isopropylacetanilide;Acilid;Albrass;Bexton;Bexton 4L;CP 31393;Kartex A;Muharicid;N-Isopropyl-2-chloroacetanilide;N-Isopropyl-α-chloroacetanilide;Niticid;Propachlore;Ramrod;Ramrod 65;Ramrod Flo;Satecid;α-Chloro-N-isopropylacetanilide
- Molecular weight:211,69
- CAS No.:1918-16-7
- Shipping conditions:RT



