Order Entry
Export
Orders LinkContactUsLinkComponent
Pencycuron

Pencycuron

Pencycuron
Catalog # EHERC15920000
Supplier:   EHRENSTORFER
Pencycuron
Catalog # EHERC15920000
Supplier:  EHRENSTORFER
CAS Number:  
Pencycuron
Pencycuron

Pencycuron

CAS Number:  
Price
67,20 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: Clc1ccc(CN(C2CCCC2)C(=O)Nc3ccccc3)cc1
  • Inchi: InChI=1S/C19H21ClN2O/c20-16-12-10-15(11-13-16)14-22(18-8-4-5-9-18)19(23)21-17-6-2-1-3-7-17/h1-3,6-7,10-13,18H,4-5,8-9,14H2,(H,21,23)
  • IUPAC: 1-[(4-chlorophenyl)methyl]-1-cyclopentyl-3-phenylurea
  • Packing group: III
Specifications

Specifications

  • Description:
    Pencycuron
  • UN No.:
    3077
  • Cat. No.:
    EHERC15920000
  • Formula:
    C₁₉H₂₁ClN₂O
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    N-[(4-Chlorophenyl)methyl]-N-cyclopentyl-N'-phenylurea;Monceraen 250SC;Monceren;NTN 19701;Pencycuron
  • Molecular weight:
    328,84
  • CAS No.:
    66063-05-6
  • Shipping conditions:
    RT

Specifications