PCB No. 31
PCB No. 31
Catalog # EHERC20003100
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Clc1ccc(cc1)c2cc(Cl)ccc2Cl
- Inchi: InChI=1S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-7-10(14)5-6-12(11)15/h1-7H
- IUPAC: 1,4-dichloro-2-(4-chlorophenyl)benzene
- Packing group: II
Specifications
Specifications
- Description:PCB No. 31
- UN No.:3432
- Cat. No.:EHERC20003100
- Formula:C₁₂H₇Cl₃
- Storage conditions:20 °C
- Hazard class description:GHS08, GHS09
- GHS signal:Danger
- Synonym(s):1,1'-Biphenyl;2,4',5-trichloro-;Biphenyl;2,4',5-trichloro- (8CI);2,4',5-Trichloro-1,1'-biphenyl;2,4',5-Trichlorobiphenyl;4,2',5'-Trichlorobiphenyl;Delor 103;PCB 31;2,4',5-TriCB
- Molecular weight:257,54
- CAS No.:16606-02-3
- Shipping conditions:RT



