PCB No. 18
PCB No. 18
Catalog # EHERC20001800
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Clc1ccc(Cl)c(c1)c2ccccc2Cl
- Inchi: InChI=1S/C12H7Cl3/c13-8-5-6-12(15)10(7-8)9-3-1-2-4-11(9)14/h1-7H
- IUPAC: 1,4-dichloro-2-(2-chlorophenyl)benzene
- Packing group: II
Specifications
Specifications
- Description:PCB No. 18
- UN No.:3432
- Cat. No.:EHERC20001800
- Formula:C₁₂H₇Cl₃
- Storage conditions:20 °C
- Hazard class description:GHS08, GHS09
- GHS signal:Danger
- Synonym(s):1,1'-Biphenyl;2,2',5-trichloro-;2,2',5-Trichloro-1,1'-biphenyl;2,2',5-Trichlorobiphenyl;2,2',5'-Trichlorobiphenyl;2,5,2'-Trichlorobiphenyl;CB 18;CB 18 (chlorocarbon);PCB 18;2,2',5-TriCB
- Molecular weight:257,54
- CAS No.:37680-65-2
- Shipping conditions:RT



