Mycophenolate mofetil
Mycophenolate mofetil
Catalog # EHERC15390900
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)OCCN3CCOCC3
- Inchi: InChI=1S/C23H31NO7/c1-15(5-7-19(25)30-13-10-24-8-11-29-12-9-24)4-6-17-21(26)20-18(14-31-23(20)27)16(2)22(17)28-3/h4,26H,5-14H2,1-3H3/b15-4+
- IUPAC: 2-morpholin-4-ylethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
- Packing group: III
Specifications
Specifications
- Description:Mycophenolate mofetil
- UN No.:2811
- Cat. No.:EHERC15390900
- Formula:C₂₃H₃₁NO₇
- Storage conditions:4 °C
- Hazard class description:GHS06, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Mycophenolate Mofetil;Mycophenolate Mofetil
- Molecular weight:433,49
- CAS No.:128794-94-5
- Shipping conditions:20 °C



