Miconazole
Miconazole
Catalog # EHERC15260090
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1
- Inchi: InChI=1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2
- IUPAC: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole
Specifications
Specifications
- Description:Miconazole
- Cat. No.:EHERC15260090
- Formula:C₁₈H₁₄Cl₄N₂O
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Miconazole;1H-Imidazole;1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-;Daktanol;Daktarin IV;Florid-F;Lauriad;MJR 1762;Monistat IV;NSC 170986;R 18134;Zimybase;Imidazole;1-[2,4-dichloro-beta-[(2,4-dichlorobenzyl)oxy]phenethyl]- (8CI);1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole;(+/-)-Miconazole;Loramyc;Isoconazole Impurity D
- Molecular weight:416,13
- CAS No.:22916-47-8
- Shipping conditions:RT



