Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol

Methanol

Methanol
Supplier:  EHRENSTORFER
Certificates
Methanol

2 Options Available Below

Product Details & Documents
  • SMILES: CO
  • Inchi: InChI=1S/CH4O/c1-2/h2H,1H3
  • IUPAC: methanol
  • Packing group: II

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • EHERC14995000
  • EHERCA14995000
  • Synonym(s):
  • Methanol;Methyl alcohol
  • Methanol;Methyl alcohol
  • CAS No.:
  • 67-56-1
  • 67-56-1
  • Formula:
  • CH₄O
  • CH₄O
  • Molecular weight:
  • 32,04
  • 32,04
  • GHS signal:
  • Danger
  • Danger
  • Hazard class description:
  • GHS02, GHS06, GHS08
  • GHS02, GHS06, GHS08
  • UN No.:
  • 1230
  • 1230
  • Shipping conditions:
  • RT
  • RT
  • Storage conditions:
  • 20 °C
  • 20 °C

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • Methanol
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 21,40 €
    21,40 €
    1 * 1 mL
       21,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    EHERC14995000
    Synonym(s)
    Methanol;Methyl alcohol
    CAS No.
    67-56-1
    Formula
    CH₄O
    Molecular weight
    32,04
    GHS signal
    Danger
    Hazard class description
    GHS02, GHS06, GHS08
    UN No.
    1230
    Shipping conditions
    RT
    Storage conditions
    20 °C
  • Methanol
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 42,70 €
    42,70 €
    1 * 1 mL
       42,70 €

    Specifications:
    More
    Specifications
    Cat. No.
    EHERCA14995000
    Synonym(s)
    Methanol;Methyl alcohol
    CAS No.
    67-56-1
    Formula
    CH₄O
    Molecular weight
    32,04
    GHS signal
    Danger
    Hazard class description
    GHS02, GHS06, GHS08
    UN No.
    1230
    Shipping conditions
    RT
    Storage conditions
    20 °C