Isovaleric acid
Isovaleric acid
Catalog # EHERC14479470
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)CC(=O)O
- Inchi: InChI=1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)
- IUPAC: 3-methylbutanoic acid
- Packing group: II
Specifications
Specifications
- Description:Isovaleric acid
- UN No.:2922
- Cat. No.:EHERC14479470
- Formula:C₅H₁₀O₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS06, GHS07
- GHS signal:Danger
- Synonym(s):Butanoic acid;3-methyl- (9CI;ACI);3-Methylbutanoic acid (ACI);Isovaleric acid (8CI);3-Methyl-1-butanoic acid;3-Methyl-n-butyric acid;3-Methylbutyrate;3-Methylbutyric acid;Acetic acid;isopropyl-;Delphinic acid;Isopentanoic acid;Isopropylacetic acid;Isovalerianic acid;NSC 62783;β-Methylbutyric acid
- Molecular weight:102,13
- CAS No.:503-74-2
- Shipping conditions:RT



