Flazasulfuron
Flazasulfuron
Catalog # EHERC13655600
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2ncccc2C(F)(F)F)n1
- Inchi: InChI=1S/C13H12F3N5O5S/c1-25-8-6-9(26-2)19-11(18-8)20-12(22)21-27(23,24)10-7(13(14,15)16)4-3-5-17-10/h3-6H,1-2H3,(H2,18,19,20,21,22)
- IUPAC: 1-(4,6-dimethoxypyrimidin-2-yl)-3-[3-(trifluoromethyl)pyridin-2-yl]sulfonylurea
- Packing group: III
Specifications
Specifications
- Description:Flazasulfuron
- UN No.:3077
- Cat. No.:EHERC13655600
- Formula:C₁₃H₁₂F₃N₅O₅S
- Storage temperature:20 °C
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):N-[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]-3-(trifluoromethyl)-2-pyridinesulfonamide;1-(4,6-Dimethoxypyrimidin-2-yl)-3-(3-trifluoromethyl-2-pyridylsulfonyl)urea;Chikara;Flazasulfuron;Katana;OK 1166;SL 160;Shibagen;Shibagen DF;2-Pyridinesulfonamide;N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-3-(trifluoromethyl)-
- Molecular weight:407,33
- CAS No.:104040-78-0
- Shipping conditions:RT



