Fenchlorphos
Fenchlorphos
Catalog # EHERC13460000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COP(=S)(OC)Oc1cc(Cl)c(Cl)cc1Cl
- Inchi: InChI=1S/C8H8Cl3O3PS/c1-12-15(16,13-2)14-8-4-6(10)5(9)3-7(8)11/h3-4H,1-2H3
- IUPAC: dimethoxy-sulfanylidene-(2,4,5-trichlorophenoxy)-λ^{5}-phosphane
- Packing group: III
Specifications
Specifications
- Description:Fenchlorphos
- UN No.:2783
- Cat. No.:EHERC13460000
- Formula:C₈H₈Cl₃O₃PS
- Storage conditions:–18 °C
- Hazard class description:GHS06, GHS07, GHS09
- GHS signal:Danger
- Synonym(s):Phosphorothioic acid;O,O-dimethyl O-(2,4,5-trichlorophenyl) ester;Blitex;Dermafos;Dow ET 14;Dow ET 57;ENT 23,284;ET 14;ET 57;Ectoral;Etrolene;Fenchlorfos;Fenchlorphos;Fenclofos;Fenclorfos;Fenclorvur;Gesektin K;Korlan;Korlane;Moorman's Medicated Rid-Ezy;NSC 8926;Nanchor;Nankor;O,O-Dimethyl O-(2,4,5-trichlorophenyl) phosphorothioate;O,O-Dimethyl O-(2,4,5-trichlorophenyl) phosphorothionate;O,O-Dimethyl O-(2,4,5-trichlorophenyl) thiophosphate;OMS 123;Phenchlorfos;Remelt;Ronnel;Rovan;Trichlormetaphos;Trichlorometaphos;Trolen;Trolene;Viozene
- Molecular weight:321,55
- CAS No.:299-84-3
- Shipping conditions:RT



