Ethylene glycol-monophenyl ether
Ethylene glycol-monophenyl ether
Catalog # EHERC13328300
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OCCOc1ccccc1
- Inchi: InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2
- IUPAC: 2-phenoxyethanol
Specifications
Specifications
- Description:Ethylene glycol-monophenyl ether
- Cat. No.:EHERC13328300
- Formula:C₈H₁₀O₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS07
- GHS signal:Warning
- Synonym(s):Phenoxyethanol;(2-Hydroxyethoxy)benzene;1-Hydroxy-2-phenoxyethane;2-Hydroxyethyl phenyl ether;2-Phenoxyethanol;2-Phenoxyethyl alcohol;Arosol;Dalpad A;Dowanol EP;Dowanol EPh;Emeressence 1160;Ethylene glycol monophenyl ether;Ethylene glycol phenyl ether;H 4644;Hisolve EPH;NSC 1864;Newpol EFP;PHE;PHE (alcohol);PHE-G;PHE-S;Phenova;Phenoxethol;Phenoxetol;Phenoxyethyl alcohol;Phenyl cellosolve;Plastilit DS 3431;Rokafenol F 1;Sepicide LD;beta-Hydroxyethyl phenyl ether;beta-Phenoxyethanol;beta-Phenoxyethyl alcohol
- Molecular weight:138,16
- CAS No.:122-99-6
- Shipping conditions:RT



