Emodin
Emodin
EHERC13117900
Jump to:
- SMILES: Cc1cc(O)c2C(=O)c3c(O)cc(O)cc3C(=O)c2c1
- Inchi: InChI=1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3
- IUPAC: 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
Specifications
Specifications
- Description:Emodin
- Cat. No.:EHERC13117900
- Formula:C₁₅H₁₀O₅
- Storage conditions:4 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):9,10-Anthracenedione;1,3,8-trihydroxy-6-methyl-;Anthraquinone;1,3,8-trihydroxy-6-methyl- (8CI);1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione;1,3,8-Trihydroxy-6-methyl-9,10-anthraquinone;1,3,8-Trihydroxy-6-methylanthraquinone;1,6,8-Trihydroxy-3-methylanthraquinone;3-Methyl-1,6,8-trihydroxyanthraquinone;4,5,7-Trihydroxy-2-methylanthraquinone;Archin;Emodin;Emodol;Frangula emodin;Frangulic acid;NSC 408120;NSC 622947;Rheum emodin;Schuttgelb
- Molecular weight:270,24
- CAS No.:518-82-1
- Shipping conditions:4 °C



