Econazole nitrate
Econazole nitrate
Catalog # EHERC13101100
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1
- Inchi: InChI=1S/C18H15Cl3N2O.HNO3/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4)
- IUPAC: 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
Specifications
Specifications
- Description:Econazole nitrate
- Cat. No.:EHERC13101100
- Formula:C₁₈H₁₅Cl₃N₂O·HNO₃
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Econazole Nitrate;1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate;Miconazole Nitrate Imp. B (EP) as Nitrate;1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole nitrate;Econazole nitrate;Ecostatin;Epi-Pevaryl;Gyno-Pevaril;Gynoryl;Ifenec;Micofugal;Micogin;NSC 243115;Palavale;Pargin;Pevaryl;R 14827;SQ 13050;Spectazole;(+/-)-1-[2-[(4-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole mononitrate;1-[2,4-Dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]imidazole mononitrate;(+/-)-Econazole nitrate;Miconazole Impurity B as Nitrate;Miconazole Nitrate Impurity B as Nitrate;Miconazole Impurity B as Nitrate
- Molecular weight:444,7
- CAS No.:24169-02-6
- Shipping conditions:RT



