Dichlorprop
Dichlorprop
Catalog # EHERC12510000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(Oc1ccc(Cl)cc1Cl)C(=O)O
- Inchi: InChI=1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)
- IUPAC: 2-(2,4-dichlorophenoxy)propanoic acid
Specifications
Specifications
- Description:Dichlorprop
- Cat. No.:EHERC12510000
- Formula:C₉H₈Cl₂O₃
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Propanoic acid;2-(2,4-dichlorophenoxy)-;Propionic acid;2-(2,4-dichlorophenoxy)- (6CI,7CI,8CI);2-(2,4-Dichlorophenoxy)propanoic acid;(±)-2,4-DP;(±)-2-(2,4-Dichlorophenoxy)propanoic acid;(±)-2-(2,4-Dichlorophenoxy)propionic acid;(±)-Dichlorprop;2,4-DCPPA;2,4-DP;2,4-Dichlorprop;2-(2,4-DP);2-(2,4-Dichlorophenoxy)propionic acid;AF 302;Canapur DP;Celatox DP;Cornox RK;DCP;Dichloroprop;Dichlorprop;Dichlorprop acid;Dicopur DP;Diprop;Hedonal DP;Herbizid DP;NSC 39624;U 46DP Fluid;α-(2,4-Dichlorophenoxy)propionic acid
- Molecular weight:235,06
- CAS No.:120-36-5
- Shipping conditions:RT



