Closantel
Closantel
Catalog # EHERC11691500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1cc(C(C#N)c2ccc(Cl)cc2)c(Cl)cc1NC(=O)c3cc(I)cc(I)c3O
- Inchi: InChI=1S/C22H14Cl2I2N2O2/c1-11-6-15(17(10-27)12-2-4-13(23)5-3-12)18(24)9-20(11)28-22(30)16-7-14(25)8-19(26)21(16)29/h2-9,17,29H,1H3,(H,28,30)
- IUPAC: N-[5-chloro-4-[(4-chlorophenyl)-cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide
- Packing group: III
Specifications
Specifications
- Description:Closantel
- UN No.:3439
- Cat. No.:EHERC11691500
- Formula:C₂₂H₁₄Cl₂I₂N₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):N-[5-chloro-4-[(4-chlorophenyl)cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide;Closantel;Clozantin;NSC 335306;Seponver;Zycloz
- Molecular weight:663,07
- CAS No.:57808-65-8
- Shipping conditions:RT



