Order Entry
Export
Orders LinkContactUsLinkComponent
Cimaterol

Cimaterol

Cimaterol
Catalog # EHERC11666350
Supplier:   EHRENSTORFER
Cimaterol
Catalog # EHERC11666350
Supplier:  EHRENSTORFER
CAS Number:  
Cimaterol
Cimaterol

Cimaterol

CAS Number:  
Price
232,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
  • SMILES: CC(C)NCC(O)c1ccc(N)c(c1)C#N
  • Inchi: InChI=1S/C12H17N3O/c1-8(2)15-7-12(16)9-3-4-11(14)10(5-9)6-13/h3-5,8,12,15-16H,7,14H2,1-2H3
  • IUPAC: 2-amino-5-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzonitrile
Specifications

Specifications

  • Description:
    Cimaterol
  • Cat. No.:
    EHERC11666350
  • Formula:
    C₁₂H₁₇N₃O
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07
  • GHS signal:
    Warning
  • Synonym(s):
    2-Amino-5-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]benzonitrile;CL 263780;Cimaterol
  • Molecular weight:
    219,28
  • CAS No.:
    54239-37-1
  • Shipping conditions:
    RT

Specifications