Carbophenothion-sulfoxide
Carbophenothion-sulfoxide
Catalog # EHERC11020220
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCOP(=S)(OCC)SCS(=O)c1ccc(Cl)cc1
- Inchi: InChI=1S/C11H16ClO3PS3/c1-3-14-16(17,15-4-2)18-9-19(13)11-7-5-10(12)6-8-11/h5-8H,3-4,9H2,1-2H3
- IUPAC: (4-chlorophenyl)sulfinylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane
- Packing group: II
Specifications
Specifications
- Description:Carbophenothion-sulfoxide
- UN No.:2810
- Cat. No.:EHERC11020220
- Formula:C₁₁H₁₆ClO₃PS₃
- Storage conditions:4 °C
- Hazard class description:GHS06, GHS09
- GHS signal:Danger
- Synonym(s):Phosphorodithioic acid;S-[[(p-chlorophenyl)sulfinyl]methyl] O,O-diethyl ester (7CI,8CI);Methanethiol;[(p-chlorophenyl)sulfinyl]-;S-ester with O,O-diethyl phosphorodithioate (8CI);Carbofenothion sulfoxide;Carbophenothion sulfoxide;R 1988;S-(p-Chlorophenylsulfinylmethyl) O,O-diethyl phosphorodithioate;Trithion sulfoxide
- Molecular weight:358,86
- CAS No.:17297-40-4
- Shipping conditions:RT



