Bromophos-ethyl
Bromophos-ethyl
Catalog # EHERC10740000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCOP(=S)(OCC)Oc1cc(Cl)c(Br)cc1Cl
- Inchi: InChI=1S/C10H12BrCl2O3PS/c1-3-14-17(18,15-4-2)16-10-6-8(12)7(11)5-9(10)13/h5-6H,3-4H2,1-2H3
- IUPAC: (4-bromo-2,5-dichlorophenoxy)-diethoxy-sulfanylidene-λ^{5}-phosphane
- Packing group: III
Specifications
Specifications
- Description:Bromophos-ethyl
- UN No.:3018
- Cat. No.:EHERC10740000
- Formula:C₁₀H₁₂BrCl₂O₃PS
- Storage conditions:–18 °C
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Bromophos A;Bromophos-ethyl;Ethyl bromophos;Filariol;Filariol 60;Nexagan;Nexagan G;O,O-Diethyl O-(2,5-dichloro-4-bromophenyl) thiophosphate;O-(4-Bromo-2,5-dichlorophenyl) O,O-diethyl phosphorothioate;OMS 659
- Molecular weight:394,05
- CAS No.:4824-78-6
- Shipping conditions:RT



