Bisphenol S-monobenzyl ether
Bisphenol S-monobenzyl ether
Catalog # EHERC10655946
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1ccc(cc1)S(=O)(=O)c2ccc(OCc3ccccc3)cc2
- Inchi: InChI=1S/C19H16O4S/c20-16-6-10-18(11-7-16)24(21,22)19-12-8-17(9-13-19)23-14-15-4-2-1-3-5-15/h1-13,20H,14H2
- IUPAC: 4-(4-benzyloxyphenyl)sulfonylphenol
- Packing group: III
Specifications
Specifications
- Description:Bisphenol S-monobenzyl ether
- UN No.:3077
- Cat. No.:EHERC10655946
- Formula:C₁₉H₁₆O₄S
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):4-[[4-(Phenylmethoxy)phenyl]sulfonyl]phenol;4-Benzyloxy-4'-hydroxydiphenyl sulfone;4-Benzyloxyphenyl 4-hydroxyphenyl sulfone;4-Benzyloxyphenyl-4'-hydroxyphenyl sulfone;4-Hydroxy-4'-benzyloxydiphenyl sulfone;BPS-BN;BPS-MA 3;p-(p-Benzyloxyphenylsulfonyl)phenol
- Molecular weight:340,39
- CAS No.:63134-33-8
- Shipping conditions:20 °C



