Benzoic acid-ethyl ester
Benzoic acid-ethyl ester
Catalog # EHERC10537750
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCOC(=O)c1ccccc1
- Inchi: InChI=1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3
- IUPAC: ethyl benzoate
Specifications
Specifications
- Description:Benzoic acid-ethyl ester
- Cat. No.:EHERC10537750
- Formula:C₉H₁₀O₂
- Storage conditions:20 °C
- Synonym(s):Benzoic acid ethyl ester;Benzoyl ethyl ether;Ethyl benzenecarboxylate;Ethyl benzoate;NSC 8884;Benzoyl Peroxide Hydrous Impurity C;Benzoyl Peroxide Impurity C
- Molecular weight:150,17
- CAS No.:93-89-0
- Shipping conditions:RT



