Aziprotryne
Aziprotryne
Catalog # EHERC10380000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CSc1nc(NC(C)C)nc(N=[N+]=[N-])n1
- Inchi: InChI=1S/C7H11N7S/c1-4(2)9-5-10-6(13-14-8)12-7(11-5)15-3/h4H,1-3H3,(H,9,10,11,12)
- IUPAC: 4-azido-6-methylsulfanyl-N-propan-2-yl-1,3,5-triazin-2-amine
Specifications
Specifications
- Description:Aziprotryne
- Cat. No.:EHERC10380000
- Formula:C₇H₁₁N₇S
- Storage conditions:20 °C
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):s-Triazine;2-azido-4-(isopropylamino)-6-(methylthio)- (7CI,8CI);4-Azido-N-(1-methylethyl)-6-(methylthio)-1,3,5-triazin-2-amine;2-Azido-4-isopropylamino-6-methylmercapto-s-triazine;2-Azido-4-isopropylamino-6-methylthio-1,3,5-triazine;2-Azido-4-isopropylamino-6-methylthio-s-triazine;2-Methylthio-4-azido-6-isopropylamino-1,3,5-triazine;Aziprotryn;Aziprotryne;Brasoran;C 7019;CIBA C 7019;Mesoranil;Mezuron
- Molecular weight:225,27
- CAS No.:4658-28-0
- Shipping conditions:RT



