Apramycin sulfate
Apramycin sulfate
Catalog # EHERC10293000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]4O[C@H](CO)[C@@H](N)[C@H](O)[C@H]4O.OS(=O)(=O)O
- Inchi: InChI=1S/C21H41N5O11.H2O4S/c1-26-11-14(30)18-8(33-20(11)37-21-16(32)13(29)10(25)9(4-27)34-21)3-7(24)19(36-18)35-17-6(23)2-5(22)12(28)15(17)31;1-5(2,3)4/h5-21,26-32H,2-4,22-25H2,1H3;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11+,12+,13+,14-,15-,16-,17-,18+,19+,20-,21-;/m1./s1
- IUPAC: (2R,3R,4S,5S,6S)-2-[[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy-4-hydroxy-3-(methylamino)-2,3,4,4a,6,7,8,8a-octahydropyrano[3,2-b]pyran-2-yl]oxy]-5-amino-6-(hydroxymethyl)oxane-3,4-diol;sulfuric acid
Specifications
Specifications
- Description:Apramycin sulfate
- Cat. No.:EHERC10293000
- Formula:C₂₁H₄₁N₅O₁₁·H₂O₄S
- Storage conditions:4 °C
- Hazard class description:GHS08
- GHS signal:Danger
- Synonym(s):Apramycin Sulfate;Apramycin Sulfate
- Molecular weight:637,66
- CAS No.:65710-07-8
- Shipping conditions:RT



