Order Entry
Export
Orders LinkContactUsLinkComponent
Aklomide

Aklomide

Aklomide
Catalog # EHERC10049000
Supplier:   EHRENSTORFER
Aklomide
Catalog # EHERC10049000
Supplier:  EHRENSTORFER
CAS Number:  
Aklomide
Aklomide

Aklomide

CAS Number:  
Price
147,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: NC(=O)c1ccc(cc1Cl)[N+](=O)[O-]
  • Inchi: InChI=1S/C7H5ClN2O3/c8-6-3-4(10(12)13)1-2-5(6)7(9)11/h1-3H,(H2,9,11)
  • IUPAC: 2-chloro-4-nitrobenzamide
Specifications

Specifications

  • Description:
    Aklomide
  • Cat. No.:
    EHERC10049000
  • Formula:
    C₇H₅ClN₂O₃
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07
  • GHS signal:
    Warning
  • Molecular weight:
    200,58
  • CAS No.:
    3011-89-0
  • Shipping conditions:
    RT

Specifications