4,4'-Dibromobenzophenone
4,4'-Dibromobenzophenone
Catalog # EHERC12220000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Brc1ccc(cc1)C(=O)c2ccc(Br)cc2
- Inchi: InChI=1S/C13H8Br2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H
- IUPAC: bis(4-bromophenyl)methanone
Specifications
Specifications
- Description:4,4'-Dibromobenzophenone
- Cat. No.:EHERC12220000
- Formula:C₁₃H₈Br₂O
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Methanone;bis(4-bromophenyl)-;Benzophenone;4,4'-dibromo- (6CI,7CI,8CI);Bis(4-bromophenyl)methanone;4,4'-Dibromobenzophenone;Bis(4-bromophenyl) ketone;Bis(p-bromophenyl) ketone;NSC 86518;p,p'-Dibromobenzophenone
- Molecular weight:340,01
- CAS No.:3988-03-2
- Shipping conditions:RT



